missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, 99+%, extra pure

Catalog No. p-3735156
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
Packaging:
Fibre drum
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC123370010 1 kg Plastic bottle
AC123370025 2.5 kg Plastic bottle
AC123370250 25 kg Fibre drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC123370010 Supplier Thermo Scientific Chemicals Supplier No. 123370010
Only null left

CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 10102-17-7
Molecular Formula Na2O3S2·5H2O
Molecular Weight (g/mol) 248.18
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48.5°C
Color Colorless to White
pH 6 to 8.4 (10% soln. at 20°C)
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 99.0 to 101% (Iodimetry)
Linear Formula Na2S2O3·5H2O
Merck Index 15, 8821
Solubility Information Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in turpentine
Formula Weight 248.18
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic bottle
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only