missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, 99.999%, (trace metal basis)
CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
$360.59 - $360.59
Chemical Identifiers
| CAS | 10102-17-7 |
|---|---|
| Molecular Formula | Na2O3S2·5H2O |
| Molecular Weight (g/mol) | 248.18 |
| InChI Key | PODWXQQNRWNDGD-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate |
| PubChem CID | 61475 |
| ChEBI | CHEBI:32150 |
| IUPAC Name | disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
| SMILES | O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC436940250
|
Thermo Scientific Chemicals
436940250 |
25 g | Glass bottle |
Each for $360.59
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 10102-17-7 | |
| 248.18 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] |
| Na2O3S2·5H2O | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate |
Specifications
| 48.5°C | |
| 10102-17-7 | |
| 25 g | |
| Na2O3S2·5H2O | |
| 15,8821 | |
| Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in turpentine | |
| O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+] | |
| 248.18 | |
| CHEBI:32150 | |
| 99.999% | |
| Glass bottle | |
| 1.7200g/mL |
| White | |
| Crystalline Powder | |
| 98.5% min. (Iodimetry) | |
| Na2S2O3·5H2O | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate | |
| 61475 | |
| 248.18 | |
| Trace Metal Basis | |
| 1.72 | |
| Sodium thiosulfate pentahydrate |
RUO – Research Use Only