missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate pentahydrate, 99.999%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| Melting Point | 48.5°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Packaging | Glass bottle |
| Specific Gravity | 1.72 |
| Assay Percent Range | 98.5% min. (Iodimetry) |
| Density | 1.7200g/mL |
| Linear Formula | Na2S2O3·5H2O |
| Merck Index | 15,8821 |
| Show More |
RUO – Research Use Only