missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiosulfate pentahydrate, 99.5+%, ACS reagent

Catalog No. AC424460051
Change view
Click to view available options
Quantity:
5 kg
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424460051 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC424460051 Supplier Thermo Scientific Chemicals Supplier No. 424460051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 10102-17-7
MDL Number MFCD00149186
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Specifications

Melting Point 48.5°C
Color White
pH 6.0 to 8.4 (5% soln. at 25°C)
Physical Form Crystals
Quantity 5 kg
Packaging Plastic bottle
Specific Gravity 1.72
Assay Percent Range 99.5 to 101% (Iodimetry)
Decomposition Information >45°C
Density 1.7200g/mL
Molecular Formula Na2O3S2·5H2O
Linear Formula Na2S2O3·5H2O
Merck Index 15, 8821
Solubility Information Solubility in water: 680g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in turpentine
Molecular Weight (g/mol) 248.18
Formula Weight 248.18
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less

RUO – Research Use Only