missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.500N ±0.002N (0.5M), LabChem™

Supplier:  LabChem LC250804

 View more versions of this product

Catalog No. LC250804


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thiosulfate
Colorless
Liquid
H10Na2O8S2
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
248.18
0.500N ±0.002N (0.5M)
Poly Bottle
10102-17-7,7732-18-5,497-19-8
12.41,87.57,0.02
4 L
Na2S2O3·5H2O
Soluble in water
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Certified
Passes Test
Sulfur compounds; Carbon dioxide

Chemical Identifiers

10102-17-7
248.17
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium pentahydrate sulfanidesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions