missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate, Certified, 0.1000N ±0.0005N (0.1M), LabChem™
Supplier: LabChem LC250606
Specifications
| Sodium Thiosulfate | |
| Colorless | |
| Liquid | |
| H10Na2O8S2 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| disodium pentahydrate sulfanidesulfonate | |
| 61475 | |
| 248.18 | |
| 0.1000N ±0.0005N (0.1M) | |
| Cubitainer™ |
| 10102-17-7,7732-18-5,497-19-8 | |
| 2.48,97.5,0.02 | |
| 10 L | |
| Na2S2O3·5H2O | |
| Soluble in water | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O | |
| 248.17 | |
| CHEBI:32150 | |
| Certified | |
| Passes Test | |
| Sulfur compounds; Carbon dioxide |
Chemical Identifiers
| 10102-17-7 | |
| 248.17 | |
| sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate | |
| CHEBI:32150 | |
| O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O |
| H10Na2O8S2 | |
| PODWXQQNRWNDGD-UHFFFAOYSA-L | |
| 61475 | |
| disodium pentahydrate sulfanidesulfonate |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature