missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

Supplier:  LabChem LC250504

 View more versions of this product

Catalog No. LC250504


Only null left
Explore available promotions

Description

Description

Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M)

Specifications

Specifications

Sodium Thiosulfate
Colorless
Liquid
H10Na2O8S2
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
248.18
0.0500N ±0.0005N (0.05M)
Poly Bottle
10102-17-7,7732-18-5,497-19-8
1.24,98.74,0.02
4 L
Na2S2O3·5H2O
Soluble in water
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Certified
Passes Test
Sulfur compounds; Carbon dioxide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions