missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

Catalog No. p-5388265
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
LC250502 1 L
LC250504 4 L
2 options

Catalog No. LC250502

Supplier: LabChem LC250502

Only null left

Sodium Thiosulfate, Certified, 0.0500N ±0.0005N (0.05M)

Specifications

Chemical Name or Material Sodium Thiosulfate
Color Colorless
CAS 10102-17-7, 7732-18-5, 497-19-8
CAS Max % 1.24, 98.74, 0.02
Physical Form Liquid
Quantity 1 L
Molecular Formula H10Na2O8S2
Linear Formula Na2S2O3·5H2O
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
Solubility Information Soluble in water
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17
PubChem CID 61475
ChEBI CHEBI:32150
Formula Weight 248.18
Grade Certified
Concentration or Composition (by Analyte or Components) 0.0500N ±0.0005N (0.05M)
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Sulfur compounds; Carbon dioxide
Show More Show Less