missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate AR Crystal, Macron Fine Chemicals™

macron

$470.67 - $470.67

Chemical Identifiers

CAS 10102-17-7
Molecular Formula H10Na2O8S2
Molecular Weight (g/mol) 248.172
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
PubChem CID 61475
ChEBI CHEBI:32150
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
SMILES O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
MK8100500
View Documents
Avantor Macron
810004
500 g POLYSTORMOR N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10102-17-7
248.172
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
CHEBI:32150
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
61475
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate

Specifications

10102-17-7
500 g
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
248.172
CHEBI:32150
POLYSTORMOR
Crystalline
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
61475
248.19
Sodium Thiosulfate, 5-Hydrate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'