missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate AR Crystal, Macron Fine Chemicals™
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 500 g |
| Formula Weight | 248.19 |
| Grade | ACS |
| Packaging | POLYSTORMOR |
| Chemical Name or Material | Sodium Thiosulfate, 5-Hydrate |