missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, Anhydrous, Granular, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390962
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004261 500 g Poly Bottle
02004262 2.5 kg Wide Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004261 Supplier Avantor Supplier No. 395301
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Physical Form Granules
Quantity 500 g
IUPAC Name disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane
Formula Weight 158.11
Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.67kg
Chemical Name or Material Sodium Thiosulfate, Anhydrous