missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Anhydrous (Certified), Fisher Chemical™

Catalog No. p-34381
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S446500 500 g Poly Bottle
S4463 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S446500 Supplier Thermo Fisher Scientific Supplier No. S446 500
Only null left

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Melting Point 48°C
Color White
pH 6.0 to 8.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥98 %
Formula Weight 158.11
Percent Purity ≥98%
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Vapor Pressure Negligible
Chemical Name or Material Sodium Thiosulfate Anhydrous
Show More Show Less