missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 1.0 N Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S5451LT

 View more versions of this product

Catalog No. 18613944

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

540-72-7,7732-18-5,10102-17-7
CNNaS
[Na+].[S-]C#N
81.07
Amber Glass Bottle
1 L
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium cyanosulfanide
1.0 N ± 0.03 N

Chemical Identifiers

540-72-7
81.07
sodium cyanosulfanide
CNNaS
VGTPCRGMBIAPIM-UHFFFAOYSA-M
[Na+].[S-]C#N
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.