missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.500 N (N/2), Ricca Chemical

$90.06 - $288.66

Specifications

Color Colorless
CAS 10102-17-7,7732-18-5,497-19-8
CAS Min % 7.67,90.0,0.06
CAS Max % 7.98,94.0,0.06
Molecular Formula H10Na2O8S2
Synonym sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
InChI Key PODWXQQNRWNDGD-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
IUPAC Name disodium pentahydrate sulfanidesulfonate
Molecular Weight (g/mol) 248.17

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
7966-16
View Documents
Ricca Chemical Company
796616
500 mL Natural Poly Bottle
Each for $90.06
Only null left
 
7966-32
View Documents
Ricca Chemical Company
796632
1 L Natural Poly Bottle
Each for $130.75
Only null left
 
7966-1
View Documents
Ricca Chemical Company
79661
4 L Natural Poly Bottle
Each for $288.66
Only null left
 
Description

Description

Specifications

Specifications

Colorless
7.67,90.0,0.06
H10Na2O8S2
PODWXQQNRWNDGD-UHFFFAOYSA-L
disodium pentahydrate sulfanidesulfonate
61475
Laboratory
∼10
Liquid
Sodium Thiosulfate
10102-17-7,7732-18-5,497-19-8
7.98,94.0,0.06
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
O.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
248.17
CHEBI:32150
Natural Poly Bottle
500 mL
0.500Normal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'