missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5390959
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Cubetainer
Narrow Mouth Polyethylene Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02004254 1 L Narrow Mouth Polyethylene Bottle
02004255 4 L Cubetainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02004254 Supplier Avantor Supplier No. 563702
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7772-98-7
Molecular Formula Na2O3S2
Molecular Weight (g/mol) 158.10
MDL Number MFCD00003499
InChI Key AKHNMLFCWUSKQB-UHFFFAOYSA-L
Synonym sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt
PubChem CID 24477
IUPAC Name disodium sulfanidesulfonate
SMILES [Na+].[Na+].[O-]S([S-])(=O)=O

Specifications

Quantity 1 L
Formula Weight 158.11
Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.01kg
Chemical Name or Material Sodium Thiosulfate, 0.1N Volumetric Solution