missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, 0.01N, Macron Fine Chemicals™
macron
$367.22 - $367.22
Chemical Identifiers
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| IUPAC Name | disodium sulfanidesulfonate |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
Chemical Identifiers
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| disodium sulfanidesulfonate |
Specifications
| White | |
| Crystals | |
| Na2O3S2 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 248.18 | |
| 1.667g/cm3 |
| 7772-98-7 | |
| 1 L | |
| MFCD00003499 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium sulfanidesulfonate | |
| 24477 | |
| narrow mouth polyethylene bottle | |
| Sodium thiosulfate |