missing translation for 'onlineSavingsMsg'
Learn More

Sodium thiomethoxide, 90+%, technical

CAS: 5188-07-8 | CH3NaS | 70.08 g/mol

Supplier:  Thermo Scientific Chemicals 469890050

 View more versions of this product

Catalog No. AC469890050

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium thiomethoxide
90+%
CH3SNa
Solubility aq. soln.: freely soluble.
[Na+].C[S-]
70.08
Technical
5188-07-8
CH3NaS
5 g
RMBAVIFYHOYIFM-UHFFFAOYSA-M
sodium methylsulfanide
70.09

Chemical Identifiers

5188-07-8
70.08
sodium methylsulfanide
CH3NaS
RMBAVIFYHOYIFM-UHFFFAOYSA-M
[Na+].C[S-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Flammable solid.

Toxic if swallowed.

Causes severe skin burns and eye damage.

GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.

No smoking.

Wear protective gloves/protective clothing/eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Immediately call a POISON CENTER or doctor/physician.

IF SWALLOWED: Rinse mouth.

Do NOT induce vomiting.

IF ON SKIN (or hair): Take off immediately all contaminated clothing.

Rinse skin with water or shower

GHS Signal Word: Danger

EINECSNumber : 225-969-9

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only