missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thioglycolate, Powder, 98%, Spectrum™ Chemical

Catalog No. 18613909
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613909 25 g Amber Glass Bottle
18613910 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18613909 Supplier Spectrum Chemical Mfg Cor Supplier No. SO20925GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 367-51-1

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.