missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Technical, Spectrum™ Chemical

Catalog No. p-7166372
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18613908 500 g Amber Glass Bottle
18613906 2.5 kg Amber Glass Bottle
18613905 12 kg Poly Pail
3 options

Catalog No. 18613908

Supplier: Spectrum Chemical Mfg Cor S1496500GM

Only null left

Chemical Identifiers

CAS 540-72-7

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.