missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate (Crystalline/Certified ACS), Fisher Chemical

Supplier:  Thermo Fisher Scientific S441500

Catalog No. S441-500



Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Thiocyanate
White
5.5 to 7.5
500 g
CNNaS
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
[Na+].[S-]C#N
81.07
CHEBI:30952
≥98%
Pass Test
287°C
540-72-7
Solid
≥98 %
MFCD00011123
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium cyanosulfanide
516871
81.07
Certified ACS
Glass Bottle

Chemical Identifiers

540-72-7
81.07
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
sodium cyanosulfanide
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952
[Na+].[S-]C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions