missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, Crystal, ACS, 98%, Spectrum™ Chemical

Catalog No. 18613899
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613899 25 g Amber Glass Bottle
18613900 125 g Amber Glass Bottle
18613901 500 g Amber Glass Bottle
18613902 2.5 kg Amber Glass Bottle
18613903 12 kg Poly Pail
18613904 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613899 Supplier Spectrum Chemical Mfg Cor Supplier No. S149525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 540-72-7

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.