missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiocyanate, ACS Reagent, Reagents

Catalog No. p-200021237
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337663 100 g
01337662 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337663 Supplier Labchem Inc Supplier No. C1327500100C2
Only null left

Sodium Thiocyanate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate
IUPAC Name sodium cyanosulfanide
SMILES [Na+].[S-]C#N

Specifications

Melting Point 310°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.295
Chemical Name or Material Sodium Thiocyanate, ACS Reagent