missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium tetrathionate dihydrate

CAS: 13721-29-4 | H4Na2O8S4 | 306.24 g/mol

Supplier:  Thermo Scientific Chemicals 461921000

 View more versions of this product

Catalog No. AC461921000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium tetrathionate dihydrate
100 g
MFCD00150868
Solubility in water: 30.6g/L.
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O
306.24
306.27
13721-29-4
H4Na2O8S4
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2
HAEPBEMBOAIUPN-UHFFFAOYSA-L
disodium dihydrate (sulfonatodisulfanyl)sulfonate
203055

Chemical Identifiers

13721-29-4
306.24
HAEPBEMBOAIUPN-UHFFFAOYSA-L
203055
O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O
H4Na2O8S4
MFCD00150868
sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2
disodium dihydrate (sulfonatodisulfanyl)sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only