accessibility menu, dialog, popup

UserName

H2OH2ONaNaOSOOSSSOOO

CAS RN 13721-29-4

IUPAC Name: disodium dihydrate (sulfonatodisulfanyl)sulfonate
Synonyms: sodium tetrathionate
Molecular Weight (g/mol): 306.24
Molecular Formula: H4Na2O8S4
InChi Key: HAEPBEMBOAIUPN-UHFFFAOYSA-L
SMILES: O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)