missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium tetrathionate dihydrate

Catalog No. AC461920250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461920250 25 g
AC461921000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461920250 Supplier Thermo Scientific Chemicals Supplier No. 461920250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13721-29-4 ∣ H4Na2O8S4 ∣ 306.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13721-29-4
Molecular Formula H4Na2O8S4
Molecular Weight (g/mol) 306.24
MDL Number MFCD00150868
InChI Key HAEPBEMBOAIUPN-UHFFFAOYSA-L
Synonym sodium tetrathionate dihydrate, tetrathionic acid, disodium salt, dihydrate, tetrathionate sodium dihydrate, disodium tetrathionate dihydrate, sodium tetrathionate, dihydrate 6ci,7ci, disodium dihydrate tetrathionate, di dithioperoxo sulfuricacid, sodium salt, hydrate 1:2:2
PubChem CID 203055
IUPAC Name disodium dihydrate (sulfonatodisulfanyl)sulfonate
SMILES O.O.[Na+].[Na+].[O-]S(=O)(=O)SSS([O-])(=O)=O

Specifications

Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Packaging Glass Bottle
Solubility Information Solubility in water: 30.6g/L.
Formula Weight 306.27
Percent Purity 98%
Grade Ungraded
Chemical Name or Material Sodium tetrathionate dihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only