missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraphenylborate 99.5+%, TCI America™

Catalog No. p-7118263
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51301G 1 g
A513010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A51301G Supplier TCI America Supplier No. A51301G
Only null left

Chemical Identifiers

CAS 143-66-8
Molecular Formula C24H20BNa
Molecular Weight (g/mol) 342.22
MDL Number MFCD00011494
InChI Key HFSRCEJMTLMDLI-UHFFFAOYSA-N
Synonym sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
PubChem CID 2723787
IUPAC Name sodium tetraphenylboranuide
SMILES [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
UN Number 2811
Formula Weight 342.22
Percent Purity ≥99.5% (W)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tetraphenylborate [Precipitation reagent for K]
EINECSNumber (3)-2553
RTECSNumber ED3362500
TSCA Yes