missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraphenylborate, ACS, 99.5%, Spectrum™ Chemical

Catalog No. p-7166370
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18613891 10 g Amber Glass Bottle
18613892 25 g Amber Glass Bottle
18613893 100 g Amber Glass Bottle
18613894 1 kg Amber Glass Bottle
18613895 5 kg Poly Pail
5 options

Catalog No. 18613891

Supplier: Spectrum Chemical Mfg Cor S149010GM

Only null left

Chemical Identifiers

CAS 143-66-8

Specifications

CAS Max % 1
Quantity 10 g
Percent Purity 99.5%
Grade ACS
Loss on Drying 0.005
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.