missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraphenylborate, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18613891
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613891 10 g Amber Glass Bottle
18613892 25 g Amber Glass Bottle
18613893 100 g Amber Glass Bottle
18613894 1 kg Amber Glass Bottle
18613895 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613891 Supplier Spectrum Chemical Mfg Cor Supplier No. S149010GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 143-66-8

Specifications

CAS Max % 1
Quantity 10 g
Percent Purity 99.5%
Grade ACS
Loss on Drying 0.005
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.