missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraphenylborate, 99%

CAS: 143-66-8 | C24H20BNa | 342.22 g/mol

Supplier:  Thermo Scientific Chemicals A1090914

 View more versions of this product

Catalog No. AAA1090914


Only null left
Explore available promotions

Description

Description

Sodium tetraphenylborate is used as a precipitating reagent for potassium and as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetraphenylborate is used as a precipitating reagent for potassium and as a protecting agent to protect amino acids. It is used in the preparation of N-acylammonium salts. It finds application as a phenyl donor in palladium-catalyzed cross-coupling reactions to prepare arylalkenes and biaryl compounds. It is used in inorganic and organometallic chemistry due to its solubility in nonpolar solvents. Furthermore, it is involved in the isolation of complexes having dinitrogen ligands.

Solubility
Soluble in water and ethanol.

Notes
Light sensitive. Incompatible with strong acids and strong oxidizing agents.
Specifications

Specifications

Sodium tetraphenylborate
>300°C
C24H20BNa
MFCD00011494
UN2811
Light sensitive
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
HFSRCEJMTLMDLI-UHFFFAOYSA-N
sodium;tetraphenylboranuide
2723787
99%
143-66-8
8
NaB(C6H5)4
25 g
3599783
14,8690
Soluble in water and ethanol.
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
342.22
342.23

Chemical Identifiers

143-66-8
342.22
HFSRCEJMTLMDLI-UHFFFAOYSA-N
2723787
C24H20BNa
MFCD00011494
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-605-5

RTECSNumber : ED3362500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only