missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Hydrate 98.0+%, TCI America™

Catalog No. p-7135890
Change view
Click to view available options
Quantity:
200 mg
1 g
Catalog No. Quantity
S0447200MG 200 mg
S04471G 1 g
2 options

Catalog No. S0447200MG

Supplier: TCI America S0447200MG

Only null left

Chemical Identifiers

CAS 79060-88-1
Molecular Formula C32H12BF24Na
Molecular Weight (g/mol) 886.209
MDL Number MFCD00043323
InChI Key LTGMONZOZHXAHO-UHFFFAOYSA-N
Synonym sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
PubChem CID 23681909
IUPAC Name sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
SMILES [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]

Specifications

Quantity 200 mg
Formula Weight 886.21
Percent Purity ≥98.0% (HPLC,W)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Tetrakis[3,5-bis(trifluoromethyl)phenyl]borate Hydrate