missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrachloroplatinate(II) hydrate, Premion™, 99.95% (metals basis), Pt 42.4% min

Catalog No. p-7048944
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1216803 1 g
AA1216806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1216803 Supplier Thermo Scientific Chemicals Supplier No. 01216803

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 207683-21-4 | Cl4Na2Pt | 382.87 g/mol

Sodium tetrachloroplatinate(II) hydrate is used as chemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207683-21-4
Molecular Formula Cl4Na2Pt
Molecular Weight (g/mol) 382.87
MDL Number MFCD00150188
InChI Key DOAIHRYZKUCMDX-UHFFFAOYSA-J
Synonym sodium tetrachloroplatinate ii hydrate, disodium tetrachloroplatinate 2-hydrate, sodium tetrachloroplatinate 2--water 2/1/1
PubChem CID 56846332
IUPAC Name disodium;tetrachloroplatinum(2-);hydrate
SMILES [Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--]

Specifications

Melting Point 100°C
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 382.88 (Anhydrous)
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 42.4% min.
Assay (metals basis)
Chemical Name or Material Sodium tetrachloroplatinate(II) hydrate
Hazard Category H302+H312+H332-H315-H317-H319-H335
Hazard Statement GHS H Statement
H300-H334-H318-H315-H317
Fatal if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 233-051-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only