accessibility menu, dialog, popup

UserName

CAS RN 207683-21-4

NaNaClClClPtCl

CAS RN 207683-21-4

IUPAC Name: disodium tetrachloride platinumdiuide
Synonyms: disodium tetrachloride platinumdiuide
Molecular Weight (g/mol): 382.87
Molecular Formula: Cl4Na2Pt
InChi Key: DOAIHRYZKUCMDX-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Cl-].[Cl-].[Cl-].[Cl-].[Pt--]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results