missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromopalladate(II), Premion™, 99.95% (metals basis), Pd 22.0% min

CAS: 50495-13-1 | Br4Na2Pd | 472.016 g/mol

Supplier:  Thermo Scientific Chemicals 04040706

 View more versions of this product

Catalog No. AA4040706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium tetrabromopalladate(II)
Powder
(metals basis)
MFCD00046199
MPLXAXBUDPGDHC-UHFFFAOYSA-J
disodium;tetrabromopalladium(2-)
16217381
99.95%
Pd 22.0% min.
50495-13-1
5 g
Br4Na2Pd
sodium tetrabromopalladate ii, br4pd.2na, disodium tetrabromopalladiumdiuide, sodium tetrabromopalladate ii , premion
[Na+].[Na+].Br[Pd-2](Br)(Br)Br
472.016
472.02
Premion

Chemical Identifiers

50495-13-1
472.016
MPLXAXBUDPGDHC-UHFFFAOYSA-J
16217381
[Na+].[Na+].Br[Pd-2](Br)(Br)Br
Br4Na2Pd
MFCD00046199
sodium tetrabromopalladate ii, br4pd.2na, disodium tetrabromopalladiumdiuide, sodium tetrabromopalladate ii , premion
disodium;tetrabromopalladium(2-)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 256-605-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only