missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetrabromopalladate(II), Premion™, 99.95% (metals basis), Pd 22.0% min

Catalog No. p-7051078
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4040703 1 g
AA4040706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4040703 Supplier Thermo Scientific Chemicals Supplier No. 04040703
Only null left

CAS: 50495-13-1 | Br4Na2Pd | 472.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50495-13-1
Molecular Formula Br4Na2Pd
Molecular Weight (g/mol) 472.016
MDL Number MFCD00046199
InChI Key MPLXAXBUDPGDHC-UHFFFAOYSA-J
Synonym sodium tetrabromopalladate ii, br4pd.2na, disodium tetrabromopalladiumdiuide, sodium tetrabromopalladate ii , premion
PubChem CID 16217381
IUPAC Name disodium;tetrabromopalladium(2-)
SMILES [Na+].[Na+].Br[Pd-2](Br)(Br)Br

Specifications

Physical Form Powder
Quantity 1 g
Formula Weight 472.02
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pd 22.0% min.
Assay (metals basis)
Chemical Name or Material Sodium tetrabromopalladate(II)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 256-605-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only