missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™

Catalog No. p-7072034
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Catalog No. Quantity
6002375 500 g
6002373 1 kg
6002376 5 kg
3 options

Catalog No. 6002375

Supplier: Honeywell Chemicals 31457500G

Only null left

Puriss. p.a., ACS Reagent, Reagent ISO, buffer substance,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 500 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium Tetraborate Decahydrate, Reag. ISO
Hazard Statement P201-P280-P308 + P313
Precautionary Statement H360FD
EINECSNumber 215-540-4
RTECSNumber VZ2275000