missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Tetraborate Decahydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Quantity | 5 kg |
| Linear Formula | Na2B4O7 · 10H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 381.37g/mol |
| Percent Purity | ≥99.5% |
| Grade | ACS/puriss. p.a. |
| Density | 1.73g/mL at 25°C |
| Chemical Name or Material | Sodium Tetraborate Decahydrate, Reag. ISO |
Safety and Handling
| Hazard Statement | P201-P280-P308 + P313 |
| Precautionary Statement | H360FD |
| EINECSNumber | 215-540-4 |
| RTECSNumber | VZ2275000 |