missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Puriss., meets analytical spec. of BP, NF, Ph. Eur., 99.0-103.0%, Honeywell Fluka™

Catalog No. p-7111746
Change view
Click to view available options
Quantity:
2.5 kg
6 x 2.5 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001910 2.5 kg Poly bottle
60044704 6 x 2.5 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001910 Supplier Honeywell Chemicals Supplier No. 116252.5KG
Only null left

Puriss., meets analytical specification of BP, NF, Ph. Eur., 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 2.5 kg
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity 99 to 103.0%
Grade puriss.
Packaging Poly bottle
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium Tetraborate Decahydrate, Meets analytical specification of Ph. Eur.BP, NF
Hazard Statement P201-P308 + P313
Precautionary Statement H360FD
EINECSNumber 215-540-4
RTECSNumber VZ2275000