missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate (Powder or Granular/NF), Fisher Chemical™

Catalog No. p-33996
Change view
Click to view available options
Quantity:
500 g
12 kg
Packaging:
Fiber Drum
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S246500 500 g Poly Bottle
S24612 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S246500 Supplier Thermo Fisher Scientific Supplier No. FLS246 500
Only null left

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7 · 10 H2O
MDL Number MFCD00149193
Synonym Sodium Biborate, Sodium Pyroborate, Borax

Specifications

Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range 99 to 105 %
Formula Weight 381.37
Percent Purity 99 to 105%
Grade NF
Identification Pass Test
Packaging Poly Bottle
Density 1.7300g/cm³
Chemical Name or Material Sodium Tetraborate Decahydrate
Show More Show Less