missing translation for 'onlineSavingsMsg'
Learn More

Sodium tetraborate decahydrate, Honeywell Fluka™

Catalog No. p-7072322
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
4 x 2.5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6003202 500 g
6003199 2.5 kg
6003203 6 x 500 g
6003201 4 x 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 6003202 Supplier Honeywell Chemicals Supplier No. S9640500G
Only null left

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1303-96-4
MDL Number MFCD00149193

Specifications

Quantity 500 g
Linear Formula Na2B4O7 · 10H2O
UN Number NONH for all modes of transport
Formula Weight 381.37g/mol
Percent Purity ≥99.5%
Density 1.73g/mL at 25°C
Chemical Name or Material Sodium tetraborate decahydrate
Hazard Statement P201-P280-P308 + P313
RTECSNumber VZ2275000