missing translation for 'onlineSavingsMsg'
Learn More

Sodium Tetraborate Decahydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. S248500
Change view
Click to view available options
Quantity:
500 g
3 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S248500 500 g Poly Bottle
S2483 3 kg Poly Pail
S24810 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S248500 Supplier Thermo Fisher Scientific Supplier No. S248 500
  • ONLINE EXCLUSIVE PRICE
    $364.50
    $174.49 / Each

    Save $190.01

    52.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $2,187.00
    $1,046.94 / Case of 6 Each

    Save $1,140.06

    52.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1303-96-4
Molecular Formula B4Na2O7 · 10 H2O
MDL Number MFCD00149193
Synonym Sodium Biborate, Sodium Pyroborate, Borax

Specifications

Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 500 g
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range 99.5 to 105.0 %
Density 1.7300 g/cm³
Molecular Weight (g/mol) 381.36
Formula Weight 381.37
Percent Purity 99.5 to 105.0%
Grade ACS
Chemical Name or Material Sodium Tetraborate Decahydrate
Show More Show Less