Learn More
Sodium tetraborate decahydrate, 99.5+%, ACS reagent
CAS: 1303-96-4 | B4Na2O7·10H2O | 381.36 g/mol
Supplier: Thermo Scientific Chemicals 419450025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium tetraborate decahydrate | |
| 1303-96-4 | |
| 9.15 to 9.20 (0.01M soln. at 25°C) | |
| 2.5 kg | |
| B4Na2O7·10H2O | |
| MFCD00149193 | |
| 15, 8726 | |
| Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol,insoluble in alcohol | |
| 381.36 | |
| ACS Reagent |
| 75.0°C | |
| White | |
| Crystalline Powder | |
| 99.5+% | |
| Na2B4O7·10H2O | |
| 04,461 | |
| Borax, Sodium borate decahydrate | |
| 381.36 | |
| ≥99.5% | |
| Plastic bottle |
Chemical Identifiers
| 1303-96-4 | |
| 381.36 | |
| Borax, Sodium borate decahydrate |
| B4Na2O7·10H2O | |
| MFCD00149193 |
Safety and Handling
GHS H Statement
May damage fertility.
May damage the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RTECSNumber : VZ2275000
TSCA : TSCA
RUO – Research Use Only