missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-Butoxide 98.0+%, TCI America™

Supplier:  TCI America S0450100G

 View more versions of this product

Catalog No. S0450100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium tert-Butoxide
270°C
C4H9NaO
100 g
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
CC(C)(C)[O-].[Na+]
96.105
96.10
Crystalline Powder
865-48-5
White-Yellow
MFCD00040575
3206
MFRIHAYPQRLWNB-UHFFFAOYSA-N
sodium;2-methylpropan-2-olate
23676156
≥98.0% (T)

Chemical Identifiers

865-48-5
96.105
MFRIHAYPQRLWNB-UHFFFAOYSA-N
23676156
CC(C)(C)[O-].[Na+]
C4H9NaO
MFCD00040575
sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
sodium;2-methylpropan-2-olate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0205

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions