accessibility menu, dialog, popup

UserName

CAS RN 865-48-5

IUPAC Name: sodium 2-methylpropan-2-olate
Synonyms: sodium tert-butoxide
Molecular Weight (g/mol): 96.11
Molecular Formula: C4H9NaO
InChi Key: MFRIHAYPQRLWNB-UHFFFAOYSA-N
SMILES: [Na+].CC(C)(C)[O-]
Molecular Weight (g/mol) 
  • (3)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)