Learn More
Sodium tert-butoxide, 99%, pure
CAS: 865-48-5 | C4H9NaO | 96.1 g/mol
Supplier: Thermo Scientific Chemicals 252550025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium tert-butoxide | |
| 180.0°C | |
| 99% | |
| C4H9NaO | |
| MFCD00040575 | |
| 16,55 | |
| Solubility in water: reacts. | |
| CC(C)(C)[O-].[Na+] | |
| 96.1 | |
| 96.1 | |
| Pure |
| 865-48-5 | |
| 12°C | |
| Plastic Drum | |
| NaOC(CH3)3 | |
| 2.5 kg | |
| sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
| MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
| sodium;2-methylpropan-2-olate | |
| 23676156 | |
| 99% |
Chemical Identifiers
| 865-48-5 | |
| 96.1 | |
| MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
| 23676156 | |
| CC(C)(C)[O-].[Na+] |
| C4H9NaO | |
| MFCD00040575 | |
| sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
| sodium;2-methylpropan-2-olate |
Safety and Handling
GHS H Statement
Self-heating; may catch fire.
Causes severe skin burns and eye damage.
Reacts violently with water.
Flammable solid.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open f
GHS Signal Word: Danger
EINECSNumber : 212-741-9
RUO – Research Use Only