Learn More
Sodium tert-butoxide, 97%
CAS: 865-48-5 | C4H9NaO | 96.105 g/mol
Supplier: Thermo Scientific Chemicals A1994236
Description
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
Specifications
| Sodium tert-butoxide | |
| 0.785 | |
| C4H9NaO | |
| MFCD00040575 | |
| UN3206 | |
| Air and moisture sensitive | |
| MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
| sodium;2-methylpropan-2-olate | |
| 23676156 | |
| 97% |
| 865-48-5 | |
| Odorless | |
| (CH3)3CONa | |
| 500 g | |
| 3654215 | |
| sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
| CC(C)(C)[O-].[Na+] | |
| 96.105 | |
| 96.11 |
Chemical Identifiers
| 865-48-5 | |
| 96.105 | |
| MFRIHAYPQRLWNB-UHFFFAOYSA-N | |
| 23676156 | |
| CC(C)(C)[O-].[Na+] |
| C4H9NaO | |
| MFCD00040575 | |
| sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide | |
| sodium;2-methylpropan-2-olate |
Safety and Handling
GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P235+P410-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H500-X
DOTInformation : Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
EINECSNumber : 212-741-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only