Learn More
Sodium tert-butoxide, 97%
Description
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.
Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.
Specifications
Specifications
| Density | 0.785 |
| Odor | Odorless |
| Linear Formula | (CH3)3CONa |
| Quantity | 2500 g |
| UN Number | UN3206 |
| Beilstein | 3654215 |
| Sensitivity | Air and moisture sensitive |
| Formula Weight | 96.11 |
| Percent Purity | 97% |
| Chemical Name or Material | Sodium tert-butoxide |
Safety and Handling
| Hazard Category | H228-H314-H500-X |
| Hazard Statement | GHS H Statement H314-H318-H228 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable solid. |
| Precautionary Statement | P210-P235+P410-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S. |
| EINECSNumber | 212-741-9 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only