missing translation for 'onlineSavingsMsg'
Learn More

Sodium tert-butoxide, 97%

Catalog No. AAA1994222
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1994222 100 g
AAA1994236 500 g
AAA199420E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1994222 Supplier Thermo Scientific Chemicals Supplier No. A1994222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 865-48-5 | C4H9NaO | 96.105 g/mol

Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tert-butoxide is used as a strong base and a non-nucleophilic base. In organic synthesis, it serves as an intermediate in various reactions like condensation, rearrangement and ring-opening. Further, it finds use in agrochemicals, pharmaceuticals, colorants, aroma chemicals, detergents and biodiesel. It acts as a catalyst in polymerization and isomerization reactions.

Notes
Air sensitive and hygroscopic. Incompatible with acids, reducing agents, oxygen, water, alcohols, chlorinated solvents and halogens.

Chemical Identifiers

CAS 865-48-5
Molecular Formula C4H9NaO
Molecular Weight (g/mol) 96.105
MDL Number MFCD00040575
InChI Key MFRIHAYPQRLWNB-UHFFFAOYSA-N
Synonym sodium tert-butoxide, sodium 2-methylpropan-2-olate, sodium t-butoxide, sodium-t-butoxide, t-butoxy sodium, tert-butoxysodium, naotbu, tbuona, 2-propanol, 2-methyl-, sodium salt, sodium-tert-butoxide
PubChem CID 23676156
IUPAC Name sodium;2-methylpropan-2-olate
SMILES CC(C)(C)[O-].[Na+]

Specifications

Density 0.785
Odor Odorless
Linear Formula (CH3)3CONa
Quantity 100 g
UN Number UN3206
Beilstein 3654215
Sensitivity Air and moisture sensitive
Formula Weight 96.11
Percent Purity 97%
Chemical Name or Material Sodium tert-butoxide
Hazard Category H228-H314-H500-X
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P235+P410-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: II; Proper Shipping Name: ALKALI METAL ALCOHOLATES, SELF-HEATING, CORROSIVE, N.O.S.
EINECSNumber 212-741-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only