missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Taurocholate, Natural, Spectrum™ Chemical
SO208, 145-42-6
Supplier: Spectrum Chemical Mfg Cor SO208100GM
Description
Spectrum™ Chemical Sodium Taurocholate, Natural is a bile acid yellow in color involved in the emulsification of fats. It appears as a sodium salt in the bile of mammals. It can be administered as a choleretic and cholagogue in medical use. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Specifications
| 145-42-6 | |
| 100 g | |
| JAJWGJBVLPIOOH-UHFFFAOYNA-M | |
| sodium 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate | |
| Reagent |
| 1 | |
| C26H44NNaO7S | |
| [Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C | |
| 537.69 | |
| Poly Bottle |
Chemical Identifiers
| 145-42-6 | |
| 537.69 | |
| sodium 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate |
| C26H44NNaO7S | |
| JAJWGJBVLPIOOH-UHFFFAOYNA-M | |
| [Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C |