missing translation for 'onlineSavingsMsg'
Learn More

Sodium Taurocholate, Natural, Spectrum™ Chemical

Catalog No. 18606878
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606878 5 g Amber Glass Bottle
18606879 25 g Poly Bottle
18606880 100 g Poly Bottle
18606881 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606878 Supplier Spectrum Chemical Mfg Cor Supplier No. SO2085GM

SO208, 145-42-6

Spectrum™ Chemical Sodium Taurocholate, Natural is a bile acid yellow in color involved in the emulsification of fats. It appears as a sodium salt in the bile of mammals. It can be administered as a choleretic and cholagogue in medical use. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 145-42-6
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.69
InChI Key JAJWGJBVLPIOOH-UHFFFAOYNA-M
IUPAC Name sodium 2-(4-{4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate
SMILES [Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CC(O)C12C

Specifications

CAS Max % 1
Quantity 5 g
Grade Reagent
Packaging Amber Glass Bottle