missing translation for 'onlineSavingsMsg'
Learn More

Sodium taurocholate hydrate, 96%

Catalog No. AAA1834614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1834614 25 g
AAA1834603 1 g
AAA1834606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1834614 Supplier Thermo Scientific Chemicals Supplier No. A1834614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium taurocholate hydrate (CAS # 145-42-6) is a bile acid and detergent used in molecular biology for protein solubilization. | CAS: 345909-26-4 | C26H44NNaO7S | 537.688 g/mol

Sodium taurocholate is a bile acid having detergent capabilities; natural surfactant. It is used for solubilization of protein. In the preparation of digestive enzymes, it is used as an emulsifier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Sodium taurocholate hydrate is a bile acid conjugate of taurine and cholic acid that acts as a detergent
  • Sodium taurocholate solubilizes fats within the digestive system
  • It is the main component of bile acid in carnivorous animals; metabolites are also common in human bile

Application

  • Sodium taurocholate hydrate can be used as an emulsifier for protein isolation

Chemical Identifiers

CAS 345909-26-4
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.688
MDL Number MFCD00150819
InChI Key JAJWGJBVLPIOOH-VXFFEJGCSA-M
Synonym sodium taurocholate, taurocholic acid sodium salt, taurocholate sodium, taurocholate sodium salt, unii-m6n3th81no, monosodium n-choloyltaurinate, m6n3th81no, monosodium taurocholate, sodium n-choloyltaurinate, monosodium taurocholic acid
PubChem CID 131632374
IUPAC Name sodium;2-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C.[Na+]

Specifications

Assay Percent Range 0.97
Quantity 25 g
Solubility Information Soluble in Water (50mg/mL- Clear Solution)
Formula Weight 537.73 (Anhydrous)
Chemical Name or Material Sodium Taurocholate Hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-653-7
RTECSNumber KI8019250
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only